Ismatec™ Extension Pump Tubing, PharMed™ BPT
Ismatec™ Extension Pump Tubing protects light-sensitive samples
Supplier:  Ismatec™ MFLX9580948
 View more versions of this product
Catalog No. 13200080
Explore available promotions
Description
Description
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Specifications
    Specifications
    Autoclavable
    2.79 mm
    100 ft.
    Ismatec pumps
    Tan
    0.1098 in.
    PharMed BPT
    30.4 cm
    2.79 mm x 30.4 m L
    Extension Pump Tubing
    Product Suggestions
    Product Suggestions