missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ ISO 7&8 Cleanroom Stainless Steel Table with Leveling Glides and Solid Top

Fixed Height Cleanroom Stainless Steel Tables with Leveling Glides have a heavy duty all Type 304 stainless steel C-frame construction.

Supplier:  Fisherbrand™ FCRT3072TUS

 View more versions of this product

Catalog No. 15050259

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • Fixed Height Stainless Steel Tables with Leveling Glides have a heavy-duty stainless-steel C-frame construction; suitable for ISO 7 & 8 Cleanroom environments
  • The work top is Type 304 stainless steel, has a brushed finish with rolled edges at the front and back, and square turndown sides providing for flush adjacent installations when required.
  • These tables are furnished at a standing height with stainless steel legs that have adjustable bullet style 1 in. wide leveling feet.
  • Optional Type 304 stainless steel undershelf.
  • Tables ship knocked down.
Specifications

Specifications

2 Years
30 in.
No
No
Table Assembly
76.2 cm
1
#4 Finish
SEFA 8
None
91.8 cm
36.125 in.
No
72 in.
182.9 cm
Not Autoclavable
Stainless Steel
Silver
Videos
SDS
Documents

Documents

Promotions

Promotions