missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
iso-Propyl Alcohol, MilliporeSigma™
Ideal for HPLC
Supplier: Technidata PX18381
Specifications
iso-Propyl Alcohol | |
67-63-0 | |
0.78g/cm3 at 20°C | |
11.85°C (53.3°F) | |
Charactristic | |
Liquid | |
C3H8O | |
UN 1219 | |
Soluble in water | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99.8% |
-88.9°C | |
Colorless | |
4 L | |
399°C (750.2°F) | |
Glass Bottle | |
Meets ACS specifications, meets reagent specifications for testing USP/NF monographs | |
CH3CHOHCH3 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1 | |
HPLC |
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II
Recommended Storage : Room Temperature