missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoamyl Nitrate 98.0+%, TCI America™
Supplier: TCI America I008825G
Specifications
Isoamyl Nitrate | |
Colorless | |
C5H11NO3 | |
25 g | |
Isopentyl Nitrate, 3-Methylbutyl Nitrate | |
CC(C)CCO[N+](=O)[O-] | |
133.147 | |
133.15 | |
Liquid |
543-87-3 | |
148°C | |
MFCD00007042 | |
1112 | |
NTHGIYFSMNNHSC-UHFFFAOYSA-N | |
3-methylbutyl nitrate | |
10985 | |
≥98.0% (GC) |
Chemical Identifiers
543-87-3 | |
133.147 | |
NTHGIYFSMNNHSC-UHFFFAOYSA-N | |
10985 | |
CC(C)CCO[N+](=O)[O-] |
C5H11NO3 | |
MFCD00007042 | |
Isopentyl Nitrate, 3-Methylbutyl Nitrate | |
3-methylbutyl nitrate |
Safety and Handling
EINECSNumber : (2)-1581
RTECSNumber : NT0187000
TSCA : Yes