Learn More
Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
$91.84 - $289.59
Chemical Identifiers
CAS | 110-46-3 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.15 |
MDL Number | MFCD00002057 |
InChI Key | OWFXIOWLTKNBAP-UHFFFAOYSA-N |
Synonym | isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i |
PubChem CID | 8053 |
ChEBI | CHEBI:2691 |
IUPAC Name | 3-methylbutyl nitrite |
SMILES | CC(C)CCON=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC165701000
|
Thermo Scientific Chemicals
165701000 |
100 mL | Glass Bottle |
Each for $91.84
|
|
||||
AC165705000
|
Thermo Scientific Chemicals
165705000 |
500 mL | Glass Bottle |
Each for $289.59
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-46-3 | |
117.15 | |
OWFXIOWLTKNBAP-UHFFFAOYSA-N | |
8053 | |
3-methylbutyl nitrite |
C5H11NO2 | |
MFCD00002057 | |
isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i | |
CHEBI:2691 | |
CC(C)CCON=O |
Specifications
110-46-3,497-19-8 | |
0.8700g/mL | |
10°C | |
96% min. (GC) | |
C5H11NO2 | |
(CH3)2CHCH2CH2ONO | |
100 mL | |
01,40; 04,270; 05,358; 06,307; 09,258 | |
15, 5168 | |
Solubility in water: practically insoluble in water. Other solubilities: miscible with alcohol,chloroform and ether | |
CC(C)CCON=O | |
117.15 | |
8053 | |
117.15 | |
Liquid |
Yellow | |
96.0°C to 99.0°C | |
Authentic | |
Glass Bottle | |
1.3840 to 1.3880 | |
MFCD00002057 | |
01, 402 | |
0.87 | |
isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i | |
OWFXIOWLTKNBAP-UHFFFAOYSA-N | |
3-methylbutyl nitrite | |
0.2% sodium carbonate | |
CHEBI:2691 | |
97% | |
Isoamyl nitrite, 97% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-770-8
RTECSNumber : NT0187500
TSCA : TSCA
RUO – Research Use Only