missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutanesulfonyl chloride, 97%, Thermo Scientific Chemicals

$154.21 - $579.36

Chemical Identifiers

CAS 35432-36-1
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.624
MDL Number MFCD07369800
InChI Key NQLNHTOCFWUYQE-UHFFFAOYSA-N
Synonym isobutanesulfonyl chloride, isobutylsulfonyl chloride, 1-propanesulfonyl chloride, 2-methyl, 2-methylpropane-1-sulfonylchloride, 2-methyl-1-propylsulfonyl chloride, 2-methyl-1-propanesulfonyl chloride, 2-methyl-propane-1-sulfonyl chloride, isobutane sulfonyl chloride, isobutane-1-sulfonylchloride, acmc-1ae06
PubChem CID 182236
IUPAC Name 2-methylpropane-1-sulfonyl chloride
SMILES CC(C)CS(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6437803
View Documents
Thermo Scientific Chemicals
H6437803
1 g
Each for $154.21
 
AAH6437806
View Documents
Thermo Scientific Chemicals
H6437806
5 g
Each for $579.36
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35432-36-1
156.624
NQLNHTOCFWUYQE-UHFFFAOYSA-N
182236
CC(C)CS(=O)(=O)Cl
C4H9ClO2S
MFCD07369800
isobutanesulfonyl chloride, isobutylsulfonyl chloride, 1-propanesulfonyl chloride, 2-methyl, 2-methylpropane-1-sulfonylchloride, 2-methyl-1-propylsulfonyl chloride, 2-methyl-1-propanesulfonyl chloride, 2-methyl-propane-1-sulfonyl chloride, isobutane sulfonyl chloride, isobutane-1-sulfonylchloride, acmc-1ae06
2-methylpropane-1-sulfonyl chloride

Specifications

35432-36-1
1.20 g/mL
C4H9ClO2S
(CH3)2CHCH2SO2Cl
1 g
1748728
isobutanesulfonyl chloride, isobutylsulfonyl chloride, 1-propanesulfonyl chloride, 2-methyl, 2-methylpropane-1-sulfonylchloride, 2-methyl-1-propylsulfonyl chloride, 2-methyl-1-propanesulfonyl chloride, 2-methyl-propane-1-sulfonyl chloride, isobutane sulfonyl chloride, isobutane-1-sulfonylchloride, acmc-1ae06
CC(C)CS(=O)(=O)Cl
156.624
156.62
Liquid
Colorless
88°C (190°F)
1.454
MFCD07369800
UN3265
Moisture sensitive
NQLNHTOCFWUYQE-UHFFFAOYSA-N
2-methylpropane-1-sulfonyl chloride
182236
97%
Isobutanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only