Learn More
Isobutyl acetate, 98%, Thermo Scientific Chemicals
$44.85 - $143.21
Chemical Identifiers
CAS | 110-19-0 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00008932 |
InChI Key | GJRQTCIYDGXPES-UHFFFAOYSA-N |
Synonym | isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate |
PubChem CID | 8038 |
ChEBI | CHEBI:50569 |
IUPAC Name | 2-methylpropyl acetate |
SMILES | CC(C)COC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA14892AK
|
Thermo Scientific Chemicals
A14892AK |
250 mL |
Each for $44.85
|
|
|||||
AAA14892AP
|
Thermo Scientific Chemicals
A14892AP |
500 mL |
Each for $57.23
|
|
|||||
AAA148920F
|
Thermo Scientific Chemicals
A148920F |
2500 mL |
Each for $143.21
|
|
|||||
Description
Isobutyl acetate is used as a solvent for lacquer, thinners, sealants, perfumes and nitrocellulose. Further, it is used as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsobutyl acetate is used as a solvent for lacquer, thinners, sealants, perfumes and nitrocellulose. Further, it is used as a flavoring agent.
Solubility
Miscible with ethanol, ether, methanol and acetone. Slightly miscible with water and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
110-19-0 | |
116.16 | |
GJRQTCIYDGXPES-UHFFFAOYSA-N | |
8038 | |
2-methylpropyl acetate |
C6H12O2 | |
MFCD00008932 | |
isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate | |
CHEBI:50569 | |
CC(C)COC(C)=O |
Specifications
110-19-0 | |
0.87 | |
18°C (64°F) | |
Fruit-like | |
1.3895 | |
250 mL | |
1741909 | |
isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate | |
GJRQTCIYDGXPES-UHFFFAOYSA-N | |
2-methylpropyl acetate | |
8038 | |
116.16 | |
Liquid |
-99°C | |
116°C to 117°C | |
98% | |
C6H12O2 | |
MFCD00008932 | |
UN1213 | |
14,5130 | |
Miscible with ethanol,ether,methanol and acetone. Slightly miscible with water and carbon tetrachloride. | |
CC(C)COC(C)=O | |
116.16 | |
CHEBI:50569 | |
98% | |
Isobutyl acetate |
Safety and Handling
GHS H Statement
H225
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H225-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYL ACETATE
EINECSNumber : 203-745-1
RTECSNumber : AI4022500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only