missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl Acetoacetate 98.0+%, TCI America™

Supplier:  TCI America A081425ML

Catalog No. A081425ML


Explore available promotions

Description

Description

Specifications

Specifications

Isobutyl Acetoacetate
Colorless
C8H14O3
25 mL
ZYXNLVMBIHVDRH-UHFFFAOYSA-N
2-methylpropyl 3-oxobutanoate
522677
≥98.0% (GC)
7779-75-1
204°C
MFCD00059349
Acetoacetic Acid Isobutyl Ester
CC(C)COC(=O)CC(C)=O
158.20
158.20
Liquid

Chemical Identifiers

7779-75-1
158.20
ZYXNLVMBIHVDRH-UHFFFAOYSA-N
522677
CC(C)COC(=O)CC(C)=O
C8H14O3
MFCD00059349
Acetoacetic Acid Isobutyl Ester
2-methylpropyl 3-oxobutanoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1475&(2)-1505

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions