missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Butyrate 98.0+%, TCI America™

$29.83 - $132.35

Chemical Identifiers

CAS 539-90-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00027134
InChI Key RGFNRWTWDWVHDD-UHFFFAOYSA-N
Synonym Butyric Acid Isobutyl Ester
PubChem CID 10885
ChEBI CHEBI:87683
IUPAC Name 2-methylpropyl butanoate
SMILES CCCC(=O)OCC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B076125ML
View Documents
TCI America
B076125ML
25 mL N/A
Only null left
 
B0761500ML
View Documents
TCI America
B0761500ML
500 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

539-90-2
144.21
RGFNRWTWDWVHDD-UHFFFAOYSA-N
10885
2-methylpropyl butanoate
C8H16O2
MFCD00027134
Butyric Acid Isobutyl Ester
CHEBI:87683
CCCC(=O)OCC(C)C

Specifications

539-90-2
157°C
MFCD00027134
3272
RGFNRWTWDWVHDD-UHFFFAOYSA-N
2-methylpropyl butanoate
10885
144.21
Liquid
Colorless
C8H16O2
25 mL
Butyric Acid Isobutyl Ester
CCCC(=O)OCC(C)C
144.21
CHEBI:87683
≥98.0% (GC)
Isobutyl Butyrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0779

RTECSNumber : ET5020000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'