missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl isobutyrate, 98%, Thermo Scientific Chemicals

$211.84 - $211.84

Chemical Identifiers

CAS 97-85-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00008916
InChI Key RXGUIWHIADMCFC-UHFFFAOYSA-N
Synonym isobutyl isobutyrate, isobutyl 2-methylpropanoate, isobutyl isobutanoate, isobutyric acid, isobutyl ester, propanoic acid, 2-methyl-, 2-methylpropyl ester, 2-methylpropyl 2-methylpropionate, 2-methylpropyl isobutyrate, isobutylester kyseliny isomaselne, fema number 2189, 2-methyl-1-propyl 2-methylpropanoate
PubChem CID 7351
IUPAC Name 2-methylpropyl 2-methylpropanoate
SMILES CC(C)COC(=O)C(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL04655AU
View Documents
Thermo Scientific Chemicals
L04655AU
1000 mL N/A
 
Description

Description

Isobutyl isobutyrate is used principally as a solvent, especially for nitrocellulose lacquers and thinners as well as coatings for plastic substrates, and high-solids coatings. The majority of end uses are in coatings (automotive, industrial, wood furniture, and graphic arts).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyl isobutyrate is used principally as a solvent, especially for nitrocellulose lacquers and thinners as well as coatings for plastic substrates, and high-solids coatings. The majority of end uses are in coatings (automotive, industrial, wood furniture, and graphic arts).

Solubility
Soluble in water (1 g/L at 20oc).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

97-85-8
144.21
RXGUIWHIADMCFC-UHFFFAOYSA-N
7351
CC(C)COC(=O)C(C)C
C8H16O2
MFCD00008916
isobutyl isobutyrate, isobutyl 2-methylpropanoate, isobutyl isobutanoate, isobutyric acid, isobutyl ester, propanoic acid, 2-methyl-, 2-methylpropyl ester, 2-methylpropyl 2-methylpropionate, 2-methylpropyl isobutyrate, isobutylester kyseliny isomaselne, fema number 2189, 2-methyl-1-propyl 2-methylpropanoate
2-methylpropyl 2-methylpropanoate

Specifications

97-85-8
0.855
37°C (98°F)
C8H16O2
MFCD00008916
UN2528
14,5145
Soluble in water (1g/L at 20oc).
CC(C)COC(=O)C(C)C
144.21
144.22
Isobutyl isobutyrate
-81°C
147°C to 149°C
Fruit-like
1.3995
1000 mL
1701355
isobutyl isobutyrate, isobutyl 2-methylpropanoate, isobutyl isobutanoate, isobutyric acid, isobutyl ester, propanoic acid, 2-methyl-, 2-methylpropyl ester, 2-methylpropyl 2-methylpropionate, 2-methylpropyl isobutyrate, isobutylester kyseliny isomaselne, fema number 2189, 2-methyl-1-propyl 2-methylpropanoate
RXGUIWHIADMCFC-UHFFFAOYSA-N
2-methylpropyl 2-methylpropanoate
7351
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P374-P380-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYL ISOBUTYRATE

EINECSNumber : 202-612-5

RTECSNumber : NQ5250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only