Learn More
Isobutyl isothiocyanate, 97%
CAS: 591-82-2 | C5H9NS | 115.194 g/mol
$66.37 - $201.25
Chemical Identifiers
| CAS | 591-82-2 |
|---|---|
| Molecular Formula | C5H9NS |
| Molecular Weight (g/mol) | 115.194 |
| MDL Number | MFCD00022062 |
| InChI Key | NSDDRJXKROCWRZ-UHFFFAOYSA-N |
| Synonym | isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci |
| PubChem CID | 68960 |
| IUPAC Name | 1-isothiocyanato-2-methylpropane |
| SMILES | CC(C)CN=C=S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1893206
|
Thermo Scientific Chemicals
A1893206 |
5 g |
Each for $66.37
|
|
|||||
|
AAA1893214
|
Thermo Scientific Chemicals
A1893214 |
25 g |
Each for $201.25
|
|
|||||
Description
It is isothiocyanate compound used for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is isothiocyanate compound used for proteomics research.
Solubility
Hydrolyzes with water.
Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Chemical Identifiers
| 591-82-2 | |
| 115.194 | |
| NSDDRJXKROCWRZ-UHFFFAOYSA-N | |
| 68960 | |
| CC(C)CN=C=S |
| C5H9NS | |
| MFCD00022062 | |
| isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci | |
| 1-isothiocyanato-2-methylpropane |
Specifications
| 591-82-2 | |
| 160°C to 162°C | |
| Pungent | |
| 1.498 | |
| 5 g | |
| 1740371 | |
| isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci | |
| NSDDRJXKROCWRZ-UHFFFAOYSA-N | |
| 1-isothiocyanato-2-methylpropane | |
| 68960 | |
| 97% |
| 0.94 | |
| 24°C (75°F) | |
| C5H9NS | |
| MFCD00022062 | |
| UN2924 | |
| Moisture sensitive | |
| Hydrolyzes with water. | |
| CC(C)CN=C=S | |
| 115.194 | |
| 115.2 | |
| Isobutyl isothiocyanate |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 209-733-2
RTECSNumber : NX8780100
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only