missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isobutyl Mercaptan 98.0+%, TCI America™
Supplier: TCI America M040425ML
Specifications
Isobutyl Mercaptan | |
Yellow | |
C4H10S | |
25 mL | |
isobutyl mercaptan, 2-methyl-1-propanethiol, isobutylmercaptan, isobutanethiol, 1-propanethiol, 2-methyl, isobutyl thiol, 1-isobutanethiol, unii-9h070ufp2x, 2-methyl propanethiol, iso-c4h9sh | |
CC(C)CS | |
90.184 | |
90.18 | |
Liquid |
513-44-0 | |
88°C | |
MFCD00004882 | |
2347 | |
BDFAOUQQXJIZDG-UHFFFAOYSA-N | |
2-methylpropane-1-thiol | |
10558 | |
≥98.0% (GC) |
Chemical Identifiers
513-44-0 | |
90.184 | |
BDFAOUQQXJIZDG-UHFFFAOYSA-N | |
10558 | |
CC(C)CS |
C4H10S | |
MFCD00004882 | |
isobutyl mercaptan, 2-methyl-1-propanethiol, isobutylmercaptan, isobutanethiol, 1-propanethiol, 2-methyl, isobutyl thiol, 1-isobutanethiol, unii-9h070ufp2x, 2-methyl propanethiol, iso-c4h9sh | |
2-methylpropane-1-thiol |
Safety and Handling
EINECSNumber : (2)-0464
RTECSNumber : TZ7630000
TSCA : Yes