Learn More
Isobutylamine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A11017AU
Description
Isobutylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsobutylamine is used as pharmaceutical intermediate.
Solubility
Soluble in water (1000 mg/ml at 25°C), ether, alcohol, acetone and benzene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications
Isobutylamine | |
-85°C | |
67°C to 69°C | |
99% | |
C4H11N | |
MFCD00008146 | |
UN1214 | |
Air Sensitive | |
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine | |
KDSNLYIMUZNERS-UHFFFAOYSA-N | |
2-methylpropan-1-amine | |
6558 | |
73.14 | |
Liquid |
78-81-9 | |
0.736 | |
−9°C (15°F) | |
Amine-like | |
1.397 | |
1000 mL | |
385626 | |
14,5132 | |
Soluble in water (1000mg/ml at 25°C),ether,alcohol,acetone and benzene. | |
CC(C)CN | |
73.139 | |
CHEBI:15997 | |
99% |
Chemical Identifiers
78-81-9 | |
73.139 | |
KDSNLYIMUZNERS-UHFFFAOYSA-N | |
6558 | |
2-methylpropan-1-amine |
C4H11N | |
MFCD00008146 | |
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine | |
CHEBI:15997 | |
CC(C)CN |
Safety and Handling
GHS H Statement
H225-H301-H314-H318
Highly flammable liquid and vapor.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYLAMINE
EINECSNumber : 201-145-4
RTECSNumber : NP9900000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only