missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutylene oxide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 348501000

Catalog No. AC348501000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Isobutylene oxide
558-30-5
51.0°C to 52.0°C
Authentic
Glass bottle
1.3710 to 1.3730
100mL
0.808
Solubility in water: 30g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether
CC1(CO1)C
72.11
0.513 mm2/s (20°C)
98%
98%
0.8080g/mL
−32°C
97.5% min. (GC)
C4H8O
MFCD00066354
17, V,1, 58
isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
GELKGHVAFRCJNA-UHFFFAOYSA-N
2,2-dimethyloxirane
11208
72.11
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat

GHS Signal Word: Danger

EINECSNumber : 209-193-8

RTECSNumber : TZ3580000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only