missing translation for 'onlineSavingsMsg'
Learn More

Isobutylene Sulfide 98.0+%, TCI America™

$238.60 - $693.43

Chemical Identifiers

CAS 3772-13-2
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00191662
InChI Key HGJOFJDIHKHKAU-UHFFFAOYSA-N
Synonym 2,2-Dimethylethylene Sulfide, 2,2-Dimethylthiirane, 2-Methylpropylene Sulfide
PubChem CID 138050
IUPAC Name 2,2-dimethylthiirane
SMILES CC1(C)CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04615ML
View Documents
TCI America
I04615ML
5 mL N/A
Only null left
 
I046125ML
View Documents
TCI America
I046125ML
25 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3772-13-2
88.17
HGJOFJDIHKHKAU-UHFFFAOYSA-N
138050
CC1(C)CS1
C4H8S
MFCD00191662
2,2-Dimethylethylene Sulfide, 2,2-Dimethylthiirane, 2-Methylpropylene Sulfide
2,2-dimethylthiirane

Specifications

3772-13-2
86°C
MFCD00191662
1993
HGJOFJDIHKHKAU-UHFFFAOYSA-N
2,2-dimethylthiirane
138050
≥98.0% (GC)
Isobutylene Sulfide
Yellow
C4H8S
5 mL
2,2-Dimethylethylene Sulfide, 2,2-Dimethylthiirane, 2-Methylpropylene Sulfide
CC1(C)CS1
88.17
88.17
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'