missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 427391000

 View more versions of this product

Catalog No. AC427391000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Isobutylmagnesium bromide
926-62-5,60-29-7
34.6°C
AcroSeal™ Glass bottle
(CH3)2CHCH2MgBr
100mL
isobutylmagnesium bromide, grignard reagent, bromo 2-methylpropyl magnesium, isobutylmagnesiumbromide, bromo isobutyl magnesium, bromo isobutyl magnesium, isobutyl magnesium bromide, isobutyl magnesium bromide, iso-butyl magnesium bromide, 2-methylpropylmagnesiumbromide, 2-methylpropylmagnesiumbromide
CC(C)[CH2-].[Mg+2].[Br-]
161.32
161.32
2M solution in diethyl ether
0.9410g/mL
−34°C
C4H9BrMg
MFCD00075126
0.941
DQDWATOXYCARFV-UHFFFAOYSA-M
magnesium;2-methanidylpropane;bromide
4285172

Chemical Identifiers

926-62-5
161.32
DQDWATOXYCARFV-UHFFFAOYSA-M
4285172
CC(C)[CH2-].[Mg+2].[Br-]
C4H9BrMg
MFCD00075126
isobutylmagnesium bromide, grignard reagent, bromo 2-methylpropyl magnesium, isobutylmagnesiumbromide, bromo isobutyl magnesium, bromo isobutyl magnesium, isobutyl magnesium bromide, isobutyl magnesium bromide, iso-butyl magnesium bromide, 2-methylpropylmagnesiumbromide, 2-methylpropylmagnesiumbromide
magnesium;2-methanidylpropane;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Harmful if swallowed. Extremely flammable liquid and vapor. In contact with water releases flammable gas. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only