Learn More
Isobutylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 427398000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Isobutylmagnesium bromide | |
926-62-5,60-29-7 | |
34.6°C | |
AcroSeal™ Glass bottle | |
(CH3)2CHCH2MgBr | |
800mL | |
isobutylmagnesium bromide, grignard reagent, bromo 2-methylpropyl magnesium, isobutylmagnesiumbromide, bromo isobutyl magnesium, iso-butymagnesium bromide, iso-butymagnesium bromide, isobutyl-magnesium bromide, isobutyl-magnesium bromide, iso-butyl magnesium bromide, 2-methylpropylmagnesiumbromide | |
CC(C)[CH2-].[Mg+2].[Br-] | |
161.32 | |
161.32 |
2M solution in diethyl ether | |
0.9410g/mL | |
−34°C | |
C4H9BrMg | |
MFCD00075126 | |
0.941 | |
DQDWATOXYCARFV-UHFFFAOYSA-M | |
magnesium;2-methanidylpropane;bromide | |
4285172 |
Chemical Identifiers
926-62-5 | |
161.32 | |
DQDWATOXYCARFV-UHFFFAOYSA-M | |
4285172 | |
CC(C)[CH2-].[Mg+2].[Br-] |
C4H9BrMg | |
MFCD00075126 | |
isobutylmagnesium bromide, grignard reagent, bromo 2-methylpropyl magnesium, isobutylmagnesiumbromide, bromo isobutyl magnesium, iso-butymagnesium bromide, iso-butymagnesium bromide, isobutyl-magnesium bromide, isobutyl-magnesium bromide, iso-butyl magnesium bromide, 2-methylpropylmagnesiumbromide | |
magnesium;2-methanidylpropane;bromide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Harmful if swallowed. Extremely flammable liquid and vapor. In contact with water releases flammable gas. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.
GHS Signal Word: Danger
RUO â Research Use Only