Learn More
Isobutyryl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2447222
Description
Isobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsobutyryl chloride is used in the preparation of fully isobutyramide terminated hyperbranched polyethylenimines. It plays an important role in the separation and identification of shikonin and its ester derivatives in the root epidermis of Echium italicum L. It acts as an acylating agent for the ketimine derivatives of alfa-amino esters, nucleosides and pyrroles. Further, it is used in the preparation of organic peroxides. In addition to this, it is used as an intermediate for dyes, textile auxiliaries and peroxide compounds.
Solubility
Miscible with chloroform, glacial acetic acid, ether, toluene, methylene chloride and benzene.
Notes
Moisture sensitive. Incompatible with bases, water, strong oxidizing agents and alcohols.
Specifications
Isobutyryl chloride | |
∽-90°C | |
91°C to 93°C | |
0.98 | |
1.407 | |
100 g | |
605393 | |
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride | |
DGMOBVGABMBZSB-UHFFFAOYSA-N | |
2-methylpropanoyl chloride | |
62325 | |
98% |
79-30-1 | |
1.017 | |
8°C (46°F) | |
C4H7ClO | |
MFCD00000717 | |
UN2395 | |
Moisture sensitive | |
Miscible with chloroform,glacial acetic acid,ether,toluene,methylene chloride and benzene. | |
CC(C)C(=O)Cl | |
106.549 | |
106.55 | |
Liquid |
Chemical Identifiers
79-30-1 | |
106.549 | |
DGMOBVGABMBZSB-UHFFFAOYSA-N | |
62325 | |
CC(C)C(=O)Cl |
C4H7ClO | |
MFCD00000717 | |
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride | |
2-methylpropanoyl chloride |
Safety and Handling
GHS H Statement
H225-H314
P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
H225-H290-H302-H314-H330
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYRYL CHLORIDE
EINECSNumber : 201-194-1
RTECSNumber : UC3944000
TSCA : Yes
storageNote1 : Moisture Sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only