missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyryl chloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 160660010

Catalog No. AC160660010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Isobutyryl chloride
79-30-1
91.0°C to 93.0°C
Authentic
Glass bottle
1.4060 to 1.4080
MFCD00000717
02, 293
isobutyryl chloride, 2-methylpropionyl chloride, propanoyl chloride, 2-methyl, isobutanoyl chloride, isobutyric acid chloride, isobutyroyl chloride, isobutyrylchloride, dimethylacetyl chloride, chloro isopropyl ketone, alpha-methylpropionyl chloride
DGMOBVGABMBZSB-UHFFFAOYSA-N
2-methylpropanoyl chloride
62325
106.55
98%
1.0170g/mL
1°C
97.5% min. (GC)
C4H7ClO
(CH3)2CHCOCl
1L
1.017
Solubility in water: reacts. Other solubilities: reacts with alcohol,soluble in ether
CC(C)C(=O)Cl
106.55
0.55 mPa.s (20°C)
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if swallowed.
Fatal if inhaled.
May be corrosive to metals.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 201-194-1

RTECSNumber : UC3944000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only