missing translation for 'onlineSavingsMsg'
Learn More

(Isocyanoimino)triphenylphosphorane 95.0+%, TCI America™

$374.68 - $1124.03

Chemical Identifiers

CAS 73789-56-7
Molecular Formula C19H15N2P
Molecular Weight (g/mol) 302.32
MDL Number MFCD09038528
InChI Key NIDTXBFHPXMXTR-UHFFFAOYSA-N
Synonym isocyanoimino triphenylphosphorane, isocyanoiminotriphenylphosphorane, isocyano triphenyl-??-phosphanylidene amine, cnnpph3;, ncnpph3;, ph3pnnc;, acmc-209os9, cn-n=pph3;, n-isocyaniminotriphenylphosphorane, n-isocyanoiminotriphenylphosphorane
PubChem CID 11266621
IUPAC Name isocyano(triphenyl-λ⁵-phosphanylidene)amine
SMILES [C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I06435G
View Documents
TCI America
I06435G
5 g
Each for $374.68
Only null left
 
I064325G
View Documents
TCI America
I064325G
25 g
Each for $1,124.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

73789-56-7
302.32
NIDTXBFHPXMXTR-UHFFFAOYSA-N
11266621
[C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C19H15N2P
MFCD09038528
isocyanoimino triphenylphosphorane, isocyanoiminotriphenylphosphorane, isocyano triphenyl-??-phosphanylidene amine, cnnpph3;, ncnpph3;, ph3pnnc;, acmc-209os9, cn-n=pph3;, n-isocyaniminotriphenylphosphorane, n-isocyanoiminotriphenylphosphorane
isocyano(triphenyl-λ⁵-phosphanylidene)amine

Specifications

73789-56-7
C19H15N2P
5 g
NIDTXBFHPXMXTR-UHFFFAOYSA-N
isocyano(triphenyl-λ⁵-phosphanylidene)amine
11266621
≥95.0% (HPLC)
(Isocyanoimino)triphenylphosphorane
152°C
MFCD09038528
isocyanoimino triphenylphosphorane, isocyanoiminotriphenylphosphorane, isocyano triphenyl-??-phosphanylidene amine, cnnpph3;, ncnpph3;, ph3pnnc;, acmc-209os9, cn-n=pph3;, n-isocyaniminotriphenylphosphorane, n-isocyanoiminotriphenylphosphorane
[C-]#[N+]N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
302.32
302.32
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'