missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isododecane, mixture of isomers, tech. 80%, Thermo Scientific Chemicals

$145.52 - $480.21

Chemical Identifiers

CAS 31807-55-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00036106
InChI Key GTJOHISYCKPIMT-UHFFFAOYSA-N
Synonym isododecane, undecane, 2-methyl, 2-methylhendecane, unii-h40fl8477b, isodo-decane, acmc-1bc2q, isododecane, mixture of isomers
PubChem CID 23459
IUPAC Name 2-methylundecane
SMILES CCCCCCCCCC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA14235AP
View Documents
Thermo Scientific Chemicals
A14235AP
500 mL
Each for $145.52
 
AAA142350F
View Documents
Thermo Scientific Chemicals
A142350F
2500 mL
Each for $480.21
 
Description

Description

Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

31807-55-3
170.34
GTJOHISYCKPIMT-UHFFFAOYSA-N
23459
CCCCCCCCCC(C)C
C12H26
MFCD00036106
isododecane, undecane, 2-methyl, 2-methylhendecane, unii-h40fl8477b, isodo-decane, acmc-1bc2q, isododecane, mixture of isomers
2-methylundecane

Specifications

31807-55-3
170°C to 195°C
80%
1.421
500 mL
isododecane, undecane, 2-methyl, 2-methylhendecane, unii-h40fl8477b, isodo-decane, acmc-1bc2q, isododecane, mixture of isomers
GTJOHISYCKPIMT-UHFFFAOYSA-N
2-methylundecane
23459
80%
Liquid
0.74
42°C (108°F)
C12H26
MFCD00036106
UN3295
Immiscible with water.
CCCCCCCCCC(C)C
170.34
170.34
Technical
Isododecane, Mixture of Isomers, mixture of isomers
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H226-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 250-816-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only