Stoelting™ Isoflurane Vaporizers
Stoelting™ Isoflurane Vaporizers are used on the Gas Anesthesia Systems to ensure that the output concentration is not affected by flow rate, temperature variation, duration of use, liquid level, or back pressure fluctuation.
$7288.57 - $7288.57
Catalog Number | Mfr. No. | Model | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | For Use With (Equipment) | Price | Quantity | ||||
10-000-929
|
Stoelting™
53903 |
Standard funnel fill | Link 7 only |
Each for $7,288.57
|
|
||||
Description
- Specifically calibrated for use with LINK 7
- Extended capacity
- 10 year warranty contingent upon a 2 year service cycle
- Larger capacity for liquid anesthetic means filling the vaporizer less often which, in turn, reduces exposure to anesthetic gases.
- Available with either funnel fill or pin index/key fill systems.
Specifications
Neuroscience | |
250 mL |