missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isohexane, for pesticide residue analysis, contains <5% n-Hexane, Thermo Scientific Chemicals

$203.20 - $322.62

Chemical Identifiers

CAS 73513-42-5
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009406
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
IUPAC Name 2-methylpentane
SMILES CCCC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC383820010
SDS View Documents
Thermo Scientific Chemicals
383820010
1 L Glass Bottle
Each for $203.20
 
AC383820025
SDS View Documents
Thermo Scientific Chemicals
383820025
2.5 L Glass Bottle
Each for $322.62
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
Specifications

Chemical Identifiers

73513-42-5
86.18
AFABGHUZZDYHJO-UHFFFAOYSA-N
7892
CCCC(C)C
C6H14
MFCD00009406
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
2-methylpentane

Specifications

73513-42-5
0.6600g/mL
−23°C
C6H14
CH3CH2CH2CH(CH3)2
1 L
0.66
Solubility in water: insoluble. Other solubilities: soluble in ethanol
CCCC(C)C
86.18
86.18
Liquid
-154.0°C
60.0°C
Glass Bottle
1.3700 to 1.3720
MFCD00009406
2ppm max.
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
AFABGHUZZDYHJO-UHFFFAOYSA-N
2-methylpentane
7892
For pesticide residue analysis
Isohexane, for residue and pesticides analysis, >5% n-Hexane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only