missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Isolator Wrap™ Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 Contact Plates
Obtain fast and accurate results with a contact plate designed for environmental monitoring.
$327.37 - $327.37
Specifications
Format | 55mm Contact Plate |
---|---|
Manufacturing Origin | United States |
Media Type | Tryptic Soy Agar |
Product Type | Agar |
Includes | Lecithin, Polysorbate 80 |
Catalog Number | Mfr. No. | Quantity | Format | Media Type | Includes | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Format | Media Type | Includes | Price | Quantity | ||
R111820
|
Thermo Scientific™
NRE4782 |
10/Pk. | 55mm Contact Plate | Tryptic Soy Agar | Lecithin, Polysorbate 80 |
Pack of 10 Each for $327.37
|
|
||
Description
Effortlessly sample your environment using Thermo Scientific™ Isolator Wrap™ Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 Contact Plates.
Isolator Wrap plates are for environmental sampling. These products are manufactured in a cleanroom environment and validated sterile by gamma radiation. Contact plates use GripFit™ technology, which ensures the lid remains in place during handling. Plates are shrink-wrapped and contained in a protective outer barrier wrap, which is impermeable to isolator decontamination procedures but maintains the integrity of the medium inside. An extra sterile bag is provided for transporting the plates to the laboratory for testing.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
55mm Contact Plate | |
Tryptic Soy Agar | |
Lecithin, Polysorbate 80 |
United States | |
Agar | |
Flow Wrapped - Single Layer and Extra Sterile Bag |