missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Isomaltotriose, 97%
$230.10 - $977.00
Chemical Identifiers
CAS | 3371-50-4 |
---|---|
Molecular Formula | C18H32O16 |
Molecular Weight (g/mol) | 504.438 |
MDL Number | MFCD00070623 |
InChI Key | FBJQEBRMDXPWNX-IGSOQDMUSA-N |
Synonym | isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol |
PubChem CID | 133126476 |
IUPAC Name | (3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol |
SMILES | C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64186MC
|
Thermo Scientific Chemicals
J64186MC |
100 mg |
Each for $230.10
|
|
|||||
AAJ64186ME
|
Thermo Scientific Chemicals
J64186ME |
500 mg |
Each for $977.00
|
|
|||||
Description
Isomaltotriose is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsomaltotriose is used as pharmaceutical intermediate.
Solubility
Soluble in water
Notes
Air sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
3371-50-4 | |
504.438 | |
FBJQEBRMDXPWNX-IGSOQDMUSA-N | |
133126476 | |
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O |
C18H32O16 | |
MFCD00070623 | |
isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol | |
(3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol |
Specifications
3371-50-4 | |
C18H32O16 | |
100 mg | |
Soluble in water | |
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O | |
504.438 | |
504.44 | |
Powder |
White | |
MFCD00070623 | |
isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol | |
FBJQEBRMDXPWNX-IGSOQDMUSA-N | |
(3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol | |
133126476 | |
97% | |
Isomaltotriose |
Safety and Handling
EINECSNumber : 222-149-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only