Learn More
![](http://beta-static.fishersci.ca/images/Solvent.gif)
![](http://beta-static.fishersci.ca/images/Red.gif)
Isooctane (Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical O2994
Specifications
Isooctane | |
540-84-1 | |
0.690g/cm³ | |
4 L | |
Glass Bottle | |
0.001% max. | |
C8H18 | |
MFCD00008943 | |
0.51 mPa/s at 22°C | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
2,2,4-trimethylpentane | |
10907 | |
114.23 | |
Certified ACS Reagent |
-107°C | |
Colorless | |
98°C | |
Pass Test | |
Liquid | |
≥99 % | |
(CH3)3CCH2CH(CH3)2 | |
51mbar at 20°C | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
CC(C)CC(C)(C)C | |
114.232 | |
CHEBI:62805 | |
≥99% |
Chemical Identifiers
540-84-1 | |
114.232 | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
10907 | |
2,2,4-trimethylpentane |
C8H18 | |
MFCD00008943 | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
CHEBI:62805 | |
CC(C)CC(C)(C)C |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Solvent.gif)
![](http://beta-static.fishersci.ca/images/Red.gif)
DANGER!
Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid