missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopentyl acetate, 99%, cont. ca 10% other isomers, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B21618AE

 View more versions of this product

Catalog No. AAB21618AE


Explore available promotions

Description

Description

It is used as a masking agent, perfuming agent, and solvent in cosmetics industry. Mainly used in flavors and because of its intensive fruity taste especially for fruit flavors such as banana, strawberry, pear, apple and cherry flavors. Traces are used in certain sweet spicy types such as vanilla flavors. Occasionally used in perfumery when a fresh-fruity note is demanded. The major application is the use in nail polish removers and nail polish as a solvent. Isopentyl acetate is a widely employed ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a masking agent, perfuming agent, and solvent in cosmetics industry. Mainly used in flavors and because of its intensive fruity taste especially for fruit flavors such as banana, strawberry, pear, apple and cherry flavors. Traces are used in certain sweet spicy types such as vanilla flavors. Occasionally used in perfumery when a fresh-fruity note is demanded. The major application is the use in nail polish removers and nail polish as a solvent. Isopentyl acetate is a widely employed ester.

Solubility
Soluble in alcohol, fixed oils, kerosene and paraffin oil. Insoluble in water, glycerin and propylene glycol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Isopentyl acetate
-78°C
142°C
Fruit-like
C7H14O2
MFCD00008946
UN1104
isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
MLFHJEHSLIIPHL-UHFFFAOYSA-N
3-methylbutyl acetate
31276
130.19
123-92-2
0.876
25°C (77°F)
cont. ≈10% other isomers
1.4
100 mL
14,5111
Soluble in alcohol,fixed oils,kerosene and paraffin oil. Insoluble in water,glycerin and propylene glycol.
CC(C)CCOC(C)=O
130.19
CHEBI:31725
99%

Chemical Identifiers

123-92-2
130.19
MLFHJEHSLIIPHL-UHFFFAOYSA-N
31276
3-methylbutyl acetate
C7H14O2
MFCD00008946
isoamyl acetate, isopentyl acetate, isopentyl ethanoate, banana oil, pear oil, 3-methylbutyl ethanoate, isoamyl ethanoate, iso-amyl acetate, 3-methyl-1-butyl acetate, i-amyl acetate
CHEBI:31725
CC(C)CCOC(C)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMYL ACETATES

EINECSNumber : 204-662-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only