missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1375936

 View more versions of this product

Catalog No. AAL1375936


Explore available promotions

Description

Description

Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

Solubility
Miscible with esters, ketones, ethers, and aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and alcohols.
Specifications

Specifications

Isophorone Diisocyanate
4098-71-9
1.061
163°C (325°F)
1.484
500 g
2726467
isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
NIMLQBUJDJZYEJ-UHFFFAOYNA-N
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
169132
222.29
mixture of isomers
-60°C
153°C (10 mmHg)
C12H18N2O2
MFCD00064956
2290
Moisture sensitive
Miscible with esters,ketones,ethers,and aromatic and aliphatic hydrocarbons.
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
222.29
CHEBI:53214
98%

Chemical Identifiers

4098-71-9
222.29
NIMLQBUJDJZYEJ-UHFFFAOYNA-N
169132
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
C12H18N2O2
MFCD00064956
isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
CHEBI:53214
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H315-H319-H317-H335-H204
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.

P260-P264b-P271-P272-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c

H315-H317-H319-H330-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOPHORONE DIISOCYANATE

EINECSNumber : 223-861-6

RTECSNumber : NQ9370000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only