missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isophthalic acid, 99%, Thermo Scientific Chemicals

$57.43 - $57.43

Chemical Identifiers

CAS 121-91-5
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00002516
InChI Key QQVIHTHCMHWDBS-UHFFFAOYSA-N
Synonym isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
PubChem CID 8496
ChEBI CHEBI:30802
IUPAC Name benzene-1,3-dicarboxylic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122655000
SDS View Documents
Thermo Scientific Chemicals
122655000
500 g Plastic bottle
Each for $57.43
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-91-5
166.13
QQVIHTHCMHWDBS-UHFFFAOYSA-N
8496
benzene-1,3-dicarboxylic acid
C8H6O4
MFCD00002516
isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
CHEBI:30802
C1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

121-91-5
White
99%
C8H6O4
MFCD00002516
09, 832
isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
QQVIHTHCMHWDBS-UHFFFAOYSA-N
benzene-1,3-dicarboxylic acid
8496
166.13
Crystalline Powder
341.0°C to 343.0°C
Authentic
Plastic bottle
C6H4(CO2H)2
500 g
15, 5243
Solubility in water: 0.01g/100mL (25 c). Other solubilities: freely soluble in alcohol and glacial acetic acid,practically insoluble in benzene and petroleum,ether
C1=CC(=CC(=C1)C(=O)O)C(=O)O
166.13
CHEBI:30802
99%
Isophthalic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-506-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only