missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

$103.92 - $1338.59

Chemical Identifiers

CAS 4191-73-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016468
InChI Key CMHMMKSPYOOVGI-UHFFFAOYSA-N
Synonym isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester
PubChem CID 20161
IUPAC Name propan-2-yl 4-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=C(O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1393009
View Documents
Thermo Scientific Chemicals
A1393009
10 g
Each for $103.92
 
AAA1393018
View Documents
Thermo Scientific Chemicals
A1393018
50 g
Each for $330.53
 
AAA1393030
View Documents
Thermo Scientific Chemicals
A1393030
250 g
Each for $1,338.59
 
Description

Description

It is used as a preservative in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a preservative in cosmetics.

Solubility
Soluble in Chloroform, Ethyl Acetate.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
Specifications

Chemical Identifiers

4191-73-5
180.20
CMHMMKSPYOOVGI-UHFFFAOYSA-N
20161
CC(C)OC(=O)C1=CC=C(O)C=C1
C10H12O3
MFCD00016468
isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester
propan-2-yl 4-hydroxybenzoate

Specifications

4191-73-5
C10H12O3
10 g
isopropyl 4-hydroxybenzoate, isopropylparaben, isopropyl p-hydroxybenzoate, isopropyl paraben, p-hydroxybenzoic acid isopropyl ester, isopropylhydroxybenzoate, unii-a6eox47qk0, p-oxybenzoesaureisopropylester, 4-hydroxybenzoic acid isopropyl ester, benzoic acid, 4-hydroxy-, 1-methylethyl ester
CMHMMKSPYOOVGI-UHFFFAOYSA-N
propan-2-yl 4-hydroxybenzoate
20161
98%
84°C to 86°C
MFCD00016468
2641564
Soluble in Chloroform,Ethyl Acetate.
CC(C)OC(=O)C1=CC=C(O)C=C1
180.20
180.21
Isopropyl 4-hydroxybenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c

H315-H317-H319-H335

EINECSNumber : 224-069-3

RTECSNumber : DH2250000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only