missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 90% (v/v), Ricca Chemical
$87.59 - $294.78
Chemical Identifiers
| CAS | 67-63-0 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
| Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
| PubChem CID | 3776 |
| ChEBI | CHEBI:17824 |
| IUPAC Name | propan-2-ol |
| SMILES | CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R4211900500
|
Ricca Chemical Company
R4211900500A |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
|
R42119001A
|
Ricca Chemical Company
R42119001A |
1 L | Natural Poly Bottle |
N/A
|
|
||||
|
R42119004A
|
Ricca Chemical Company
R42119004A |
4 L | Natural Poly Bottle |
N/A
|
|
||||
Chemical Identifiers
| 67-63-0 | |
| Mixture | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
Specifications
| 67-63-0,7732-18-5 | |
| 9.0,89.0 | |
| 82°C | |
| Natural Poly Bottle | |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 90% | |
| 90% (v/v) |
| Colorless | |
| 11.0,91.0 | |
| 500 mL | |
| Liquid | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| Mixture | |
| CHEBI:17824 | |
| Laboratory | |
| Isopropanol |
Safety and Handling
ShelfLife : 24 months
Recommended Storage : Ambient Flammable