missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isopropyl Alcohol, ACS Grade, ≥ 99.5%, LabChem™
Supplier: LabChem LC157502
Specifications
Isopropyl Alcohol | |
67-63-0 | |
100 | |
785kg/m3 | |
1 L | |
Passes Test | |
Liquid | |
785kg/m3 | |
C3H8O | |
UN1219 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.09 | |
ACS |
-88°C | |
Colorless | |
Carbon dioxide; Carbon monoxide | |
82°C | |
12°C | |
Poly (HDPE) bottle | |
≤0.001% | |
C3H8O | |
2.5316mm2/s | |
≤0.2% | |
Soluble in water | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99.5% |
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Danger
EINECSNumber : 200-661-7
Recommended Storage : Room Temperature