missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isopropyl Alcohol, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.9%, Honeywell™
Supplier: Honeywell Chemicals AH3234PC
Specifications
Isopropyl Alcohol | |
67-63-0 | |
100.0000% | |
82.3°C | |
12°C | |
C3H8O | |
UN1219 | |
635639 | |
Miscible in Water at 20°C | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
>99.9% |
ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.9% | |
0.16 AU max. at 225 nm, 0.02 AU max. at 250 nm | |
0.785 g/cm3 | |
4 x 4 L | |
Poly-coated Bottle | |
MFCD00011674 | |
0.06% max. | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.11 | |
HPLC |
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
EINECSNumber : 200-661-7