missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals

$290.68 - $290.68

Chemical Identifiers

CAS 108-20-3
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC180680025
SDS View Documents
Thermo Scientific Chemicals
180680025
2.5 L Glass bottle
Each for $290.68
Case of 4 Each for $0.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-20-3
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
7914
CC(C)OC(C)C
MFCD00008880
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
2-propan-2-yloxypropane

Specifications

108-20-3,128-37-0
-85.5°C
68°C
Authentic
Glass bottle
1.3670 to 1.369
MFCD00008880
1159
0.01% max.
15, 5258
Solubility in water: 9g/L (20°C). Other solubilities: miscible with most common organic solvents
CC(C)OC(C)C
102.18
7914
102.18
99+%
Liquid
0.005% max.
0.7200g/mL
-29°C
99% min. (GC)
C6H14O
[(CH3)2CH]2O
2.5 L
01, 362
0.72
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
2-propan-2-yloxypropane
5 to 250ppm BHT
0.38 mPa.s (25°C)
180mbar at 20°C
Extra Pure
Isopropyl ether, Stabilized with BHT
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS Signal Word: Danger

EINECSNumber : 203-560-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only