missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isopropylamine 99.0+%, TCI America™
Supplier: TCI America I0165500ML
Specifications
Isopropylamine | |
-95°C | |
34°C | |
MFCD00008082 | |
1221 | |
JJWLVOIRVHMVIS-UHFFFAOYSA-N | |
propan-2-amine | |
6363 | |
59.11 | |
Liquid |
75-31-0 | |
Colorless | |
C3H9N | |
500 mL | |
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino | |
CC(C)N | |
59.112 | |
CHEBI:15739 | |
≥99.0% (GC,T) |
Chemical Identifiers
75-31-0 | |
59.112 | |
JJWLVOIRVHMVIS-UHFFFAOYSA-N | |
6363 | |
propan-2-amine |
C3H9N | |
MFCD00008082 | |
isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino | |
CHEBI:15739 | |
CC(C)N |
Safety and Handling
EINECSNumber : (2)-0131
RTECSNumber : NT8400000
TSCA : Yes