missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropylidene malonate, 97%, Thermo Scientific Chemicals

$94.59 - $960.58

Chemical Identifiers

CAS 2033-24-1
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00006638
InChI Key GXHFUVWIGNLZSC-UHFFFAOYSA-N
Synonym meldrum's acid, isopropylidene malonate, meldrums acid, 1,3-dioxane-4,6-dione, 2,2-dimethyl, sub-isopropyl malonate, 2,2-dimethyl-1,3-dioxan-4,6-dione, cycl-isopropylidene malonate, cyclic isopropylidene malonate, malonic acid cyclic isopropylidene ester, meldrum acid
PubChem CID 16249
IUPAC Name 2,2-dimethyl-1,3-dioxane-4,6-dione
SMILES CC1(OC(=O)CC(=O)O1)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1560314
View Documents
Thermo Scientific Chemicals
A1560314
25 g
Each for $94.59
 
AAA1560322
View Documents
Thermo Scientific Chemicals
A1560322
100 g
Each for $219.14
 
AAA1560336
View Documents
Thermo Scientific Chemicals
A1560336
500 g
Each for $960.58
 
Description

Description

Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.

Solubility
Soluble in water. Slightly soluble in ethanol and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

2033-24-1
144.126
GXHFUVWIGNLZSC-UHFFFAOYSA-N
16249
CC1(OC(=O)CC(=O)O1)C
C6H8O4
MFCD00006638
meldrum's acid, isopropylidene malonate, meldrums acid, 1,3-dioxane-4,6-dione, 2,2-dimethyl, sub-isopropyl malonate, 2,2-dimethyl-1,3-dioxan-4,6-dione, cycl-isopropylidene malonate, cyclic isopropylidene malonate, malonic acid cyclic isopropylidene ester, meldrum acid
2,2-dimethyl-1,3-dioxane-4,6-dione

Specifications

2033-24-1
C6H8O4
25 g
Moisture sensitive
meldrum's acid, isopropylidene malonate, meldrums acid, 1,3-dioxane-4,6-dione, 2,2-dimethyl, sub-isopropyl malonate, 2,2-dimethyl-1,3-dioxan-4,6-dione, cycl-isopropylidene malonate, cyclic isopropylidene malonate, malonic acid cyclic isopropylidene ester, meldrum acid
GXHFUVWIGNLZSC-UHFFFAOYSA-N
2,2-dimethyl-1,3-dioxane-4,6-dione
16249
98%
92°C to 96°C
MFCD00006638
117310
14,5817
Soluble in water. Slightly soluble in ethanol and chloroform.
CC1(OC(=O)CC(=O)O1)C
144.126
144.13
Isopropylidene malonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 217-992-8

RTECSNumber : JH0800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only