missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropylmagnesium bromide, 3M in 2-MeTHF, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H54840AE

Catalog No. AAH54840AE


Explore available promotions

Description

Description

Isopropylmagnesium bromide is a Grignard reagent used in organic synthesis, especially for the introduction of iso-propyl group to carbonyl compounds. It is also used as a strong base in aprotic solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylmagnesium bromide is a Grignard reagent used in organic synthesis, especially for the introduction of iso-propyl group to carbonyl compounds. It is also used as a strong base in aprotic solvent.

Solubility
Specifications

Specifications

Isopropylmagnesium bromide
C3H7BrMg
MFCD00672110
UN3399
isopropylmagnesium bromide, grignard reagent, magnesium, bromo 1-methylethyl, isopropylmagnesiumbromide, iprmgbr, i-prmgbr, bromoisopropylmagnesium, bromoisopropylmagnesium, i-propylmagnesiumbromide, i-propylmagnesium bromide, bromo-isopropyl-magnesium
UCJIVFREPMUZDC-UHFFFAOYSA-M
magnesium;propane;bromide
101909
3M in 2-MeTHF
920-39-8
(CH3)2CHMgBr
100 mL
Air and moisture sensitive; May form a precipitate
Miscible with tetrahydorfuran and diethyl ether. Immiscible with hydrocarbons.
CC(C)[Mg]Br
147.30
147.3

Chemical Identifiers

920-39-8
147.30
UCJIVFREPMUZDC-UHFFFAOYSA-M
101909
CC(C)[Mg]Br
C3H7BrMg
MFCD00672110
isopropylmagnesium bromide, grignard reagent, magnesium, bromo 1-methylethyl, isopropylmagnesiumbromide, iprmgbr, i-prmgbr, bromoisopropylmagnesium, bromoisopropylmagnesium, i-propylmagnesiumbromide, i-propylmagnesium bromide, bromo-isopropyl-magnesium
magnesium;propane;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.

P210-P223-P231+P232-P233-P240-P243-P260-P264b-P270-P271-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P372-P374-P380-P501c

H225-H260-H302-H314-H335

DOTInformation : Hazard Class: 4.3; Packaging Group: I

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only