missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals

$278.88 - $1085.66

Chemical Identifiers

CAS 1068-55-9
Molecular Formula C3H7ClMg
Molecular Weight (g/mol) 102.84
MDL Number MFCD00000466
InChI Key GPIBKUJXKCEZOH-UHFFFAOYSA-M
Synonym isopropylmagnesium chloride, magnesium, chloro 1-methylethyl, chloro 1-methylethyl magnesium, i-prmgcl, isopropylmagnesiumchloride, chloro isopropyl magnesium, isopropyl magnesium chloride, isopropylmagnesium chloride solution, iprmgcl, ipr-mgcl
PubChem CID 101955
IUPAC Name magnesium;propane;chloride
SMILES CC(C)[Mg]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51155AE
View Documents
Thermo Scientific Chemicals
H51155AE
100 mL
Each for $278.88
 
AAH51155AP
View Documents
Thermo Scientific Chemicals
H51155AP
500 mL
Each for $1,085.66
 
Description

Description

Isopropyl magnesium chloride is used primarily to add an iso-propyl group to carbonyl groups. This reagent can also be used as a strong base in aprotic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl magnesium chloride is used primarily to add an iso-propyl group to carbonyl groups. This reagent can also be used as a strong base in aprotic solvents.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

1068-55-9
102.84
GPIBKUJXKCEZOH-UHFFFAOYSA-M
101955
CC(C)[Mg]Cl
C3H7ClMg
MFCD00000466
isopropylmagnesium chloride, magnesium, chloro 1-methylethyl, chloro 1-methylethyl magnesium, i-prmgcl, isopropylmagnesiumchloride, chloro isopropyl magnesium, isopropyl magnesium chloride, isopropylmagnesium chloride solution, iprmgcl, ipr-mgcl
magnesium;propane;chloride

Specifications

1068-55-9
(CH3)2CHMgCl
100 mL
Air and moisture sensitive
It reacts violently with water.
CC(C)[Mg]Cl
102.84
102.85
Isopropylmagnesium chloride
C3H7ClMg
MFCD00000466
UN3399
isopropylmagnesium chloride, magnesium, chloro 1-methylethyl, chloro 1-methylethyl magnesium, i-prmgcl, isopropylmagnesiumchloride, chloro isopropyl magnesium, isopropyl magnesium chloride, isopropylmagnesium chloride solution, iprmgcl, ipr-mgcl
GPIBKUJXKCEZOH-UHFFFAOYSA-M
magnesium;propane;chloride
101955
1M in MeTHF
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H260-H302-H314-H335

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only