missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoproterenol Sulfate Dihydrate 98.0+%, TCI America™
Supplier: TCI America I026125G
Specifications
Isoproterenol Sulfate Dihydrate | |
83°C | |
C22H36N2O10S | |
25 g | |
ZOLBALGTFCCTJF-UHFFFAOYSA-N | |
4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;sulfuric acid | |
8239 | |
≥98.0% (HPLC,N) |
299-95-6 | |
White | |
MFCD00013139 | |
isoprenaline sulphate, isoprenaline sulfate, aleudrin, novodrine, aleudrine sulfate, isoproterenol sulfate, dl-isoprenaline sulfate, dl-isoproterenol sulfate, d,l-isoproterenol sulfate, +--isoprenaline sulfate | |
CC(C)NCC(C1=CC(=C(C=C1)O)O)O.CC(C)NCC(C1=CC(=C(C=C1)O)O)O.OS(=O)(=O)O | |
520.594 | |
520.59 | |
Crystalline Powder |
Chemical Identifiers
299-95-6 | |
520.594 | |
ZOLBALGTFCCTJF-UHFFFAOYSA-N | |
8239 | |
CC(C)NCC(C1=CC(=C(C=C1)O)O)O.CC(C)NCC(C1=CC(=C(C=C1)O)O)O.OS(=O)(=O)O |
C22H36N2O10S | |
MFCD00013139 | |
isoprenaline sulphate, isoprenaline sulfate, aleudrin, novodrine, aleudrine sulfate, isoproterenol sulfate, dl-isoprenaline sulfate, dl-isoproterenol sulfate, d,l-isoproterenol sulfate, +--isoprenaline sulfate | |
4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;sulfuric acid |
Safety and Handling
RTECSNumber : DO2273000
TSCA : No