Learn More
Isoquinoline-4-carboxaldehyde, 97%
CAS: 22960-16-3 | C10H7NO | 157.172 g/mol
$215.37 - $215.37
Chemical Identifiers
| CAS | 22960-16-3 |
|---|---|
| Molecular Formula | C10H7NO |
| Molecular Weight (g/mol) | 157.172 |
| MDL Number | MFCD00829440 |
| InChI Key | RNQQJLYJLDQGGL-UHFFFAOYSA-N |
| Synonym | isoquinoline-4-carboxaldehyde, 4-isoquinolinecarboxaldehyde, 4-formylisoquinoline, pubchem15962, acmc-20aig4, 4-isoquinolinecarbaldehyde, ksc201k4f, 4-isoquinolinecarboxaldehyde 7ci,8ci,9ci |
| PubChem CID | 10868870 |
| IUPAC Name | isoquinoline-4-carbaldehyde |
| SMILES | C1=CC=C2C(=C1)C=NC=C2C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6178803
|
Thermo Scientific Chemicals
H6178803 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 22960-16-3 | |
| 157.172 | |
| RNQQJLYJLDQGGL-UHFFFAOYSA-N | |
| 10868870 | |
| C1=CC=C2C(=C1)C=NC=C2C=O |
| C10H7NO | |
| MFCD00829440 | |
| isoquinoline-4-carboxaldehyde, 4-isoquinolinecarboxaldehyde, 4-formylisoquinoline, pubchem15962, acmc-20aig4, 4-isoquinolinecarbaldehyde, ksc201k4f, 4-isoquinolinecarboxaldehyde 7ci,8ci,9ci | |
| isoquinoline-4-carbaldehyde |
Specifications
| 22960-16-3 | |
| C10H7NO | |
| 1 g | |
| RNQQJLYJLDQGGL-UHFFFAOYSA-N | |
| isoquinoline-4-carbaldehyde | |
| 10868870 | |
| 97% |
| 101°C to 106°C | |
| MFCD00829440 | |
| isoquinoline-4-carboxaldehyde, 4-isoquinolinecarboxaldehyde, 4-formylisoquinoline, pubchem15962, acmc-20aig4, 4-isoquinolinecarbaldehyde, ksc201k4f, 4-isoquinolinecarboxaldehyde 7ci,8ci,9ci | |
| C1=CC=C2C(=C1)C=NC=C2C=O | |
| 157.172 | |
| 157.17 | |
| Isoquinoline-4-carboxaldehyde |
Safety and Handling
GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only