missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Isotemp™ General Purpose Laboratory Freezers

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Freezers are designed for everyday laboratory needs.

$13988.78 - $18478.64

Specifications

Door Style Solid
Door Material Stainless Steel
Cabinet Material Stainless Steel
Interior Stainless Steel
Capacity (English) Shelf 100 lb.
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (English) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Price Quantity  
FBG25FSSA
View Documents
Fisherbrand™
FBG25FSSA
25 cu. ft. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FBG49FSSA
View Documents
Fisherbrand™
FBG49FSSA
49 cu. ft. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Whisper-quiet operation
  • ENERGY STAR certified
  • Green, natural refrigerants
  • Adaptive defrost
  • Digital Controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock
  • Low heat output
  • Temp Range: -12°C to -30°C
Specifications

Specifications

Solid
Stainless Steel
100 lb.
ENERGYSTAR, Prop 65, cULus
Digital Microprocesor
Adaptive Defrost
115 V
Energy Efficient
Stainless Steel
Stainless Steel
45 kg
Yes
1 Rear Access Port
Key Lock
60 Hz
R290
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'