missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.
Supplier: Fisherbrand™ FBV72RSGA
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
Specifications
72 cu. ft. | |
2°C to 10°C | |
R290 | |
3 | |
100 lb. | |
Stainless Steel | |
Gray | |
1 Rear access port | |
RSGA | |
28.5 x 72 x 61.75 in. (724 x 1829 x 1569 mm) | |
9.9 A | |
115 V 60 Hz | |
332 kg |
2038 L | |
5°C | |
Glass | |
12 Full | |
45 kg | |
cULus, ENERGYSTAR | |
Microprocessor | |
13 months parts and labor | |
Key Lock | |
32 x 75.7 x 81 in.(813 x 1924 x 2057 mm) | |
NEMA 5-15 P | |
733 lb. |