Fisherbrand™ Isotemp™ Ultra Low Temperature Freezer
Fisherbrand™ Isotemp Ultra Low Temperature Freezers combine reliability and performance with cost effective operation and innovative features. Built using an advanced technology platform, refrigeration, microprocessor controls and high quality construction
Supplier: Fisherbrand™ IUE50086FARK
Description
Features:
- Minimal temperature variation and fast door opening recovery
- Quiet operation
- Low energy and HVAC costs
- Envronmentally friendly - utilizes green refrigerants and water blown foam that are SNAP compliant
Specifications
2 Shelves of Racks | |
2 years parts and labor + additional 3 yrs. compressor parts | |
Not Available |
— | |
500 box | |
115 V |
For research only