Fisherbrand™ Isotemp™ Ultra Low Temperature Freezer
Fisherbrand™ Isotemp Ultra Low Temperature Freezers combine reliability and performance with cost effective operation and innovative features. Built using an advanced technology platform, refrigeration, microprocessor controls and high quality construction
Supplier:  Fisherbrand™ IUE50086FDRLN
 View more versions of this product
Catalog No. IUE586FDRLN
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Features:
  • Minimal temperature variation and fast door opening recovery
  • Quiet operation
  • Low energy and HVAC costs
  • Envronmentally friendly - utilizes green refrigerants and water blown foam that are SNAP compliant
Specifications
Specifications
2 Shelves of Racks
2 years parts and labor + additional 3 yrs. compressor parts
LN2 Backup
500 box
208 V
Videos
SDS
Documents
Documents
Promotions
Promotions
For research only