missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Isotemp Value Undercounter Refrigerator
Chill, freeze and retreive samples safely, simply and economically with the Fisherbrand™ Isotemp Undercounter Refrigerator.
Supplier: Fisherbrand™ FBV05RPSA
Description
- Factory tested prior to shipment
- NRTL listed
- Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
- Adjustable temperature control
- Easy to clean, durable finish
Specifications
General Purpose Refrigerator | |
155.7 L | |
Adjustable | |
Hinged | |
Right | |
Steel | |
ABS Plastic | |
2 Adjustable Feet in Front | |
Mechanical Dial | |
10 years | |
Manual | |
Key Lock | |
23.5 x 23.63 x 33.5 in. (59.69 x 60.02 x 85.1 cm) | |
0.8 A | |
115 V | |
0.31 kWh/Day | |
43 kg |
5.5 cu. ft. | |
1°C to 12°C | |
R600a hydrocarbon refrigerant | |
1 | |
3 Adjustable | |
Cyclopentane | |
cULus | |
White | |
Indoor General Storage of Non-Critical Samples and Reagents | |
12 months | |
4 in. on all sides | |
Hermetic | |
17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5 cm) | |
NEMA 5-15 P | |
60 Hz | |
95 lb. |