missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ivabradine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America I0847200MG
Specifications
Ivabradine Hydrochloride | |
140°C | |
C27H37ClN2O5 | |
200 mg | |
CN(CCCN1CCC2=CC(=C(C=C2CC1=O)OC)OC)CC3CC4=CC(=C(C=C34)OC)OC.Cl | |
505.052 | |
CHEBI:85969 | |
≥98.0% (HPLC,N) |
148849-67-6 | |
White | |
MFCD00929899 | |
HLUKNZUABFFNQS-ZMBIFBSDSA-N | |
3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl-methylamino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride | |
3045381 | |
505.05 | |
Crystalline Powder |
Chemical Identifiers
148849-67-6 | |
505.052 | |
HLUKNZUABFFNQS-ZMBIFBSDSA-N | |
CHEBI:85969 | |
CN(CCCN1CCC2=CC(=C(C=C2CC1=O)OC)OC)CC3CC4=CC(=C(C=C34)OC)OC.Cl |
C27H37ClN2O5 | |
MFCD00929899 | |
3045381 | |
3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl-methylamino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator