missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synthware™ Jacketed Vigreux Distillation Head

Sealed-on condenser, braced for strength. Vacuum adapter has an inner joint and a sealed in drip tip.

Supplier:  Synthware™ D214200

 View more versions of this product

Catalog No. 31501056


Explore available promotions

Description

Description

Specifications

Specifications

350 mm
Organic Synthesis
Borosilicate Glass
25 mm Immersion Thermometer
13.8 in.
7.9 in.
Ground Glass
325 mm
Vigreux, Jacketed
12.8 in.
1 Ea.
24/40
200 mm
SDS
Documents

Documents

Promotions

Promotions