missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Slide Holder
Provides an inexpensive way for drying excess fluids from flat surfaces. Fisherbrand™ Slide Holder is constructed of chemical-resistant HDPE.
Supplier: Fisherbrand™ HS15981CF2
Description
Chemical-resistant high-density polyethylene holder
- Convenient-to-use receptacle holds both 3 x 1 in. or 75 x 25 mm slides during evacuation and cleaning
- Light, strong, chemical-resistant high-density polyethylene holder provides an inexpensive way for drying excess fluids from flat surfaces
- 10° angle of slots facilitates clear viewing of desired slide
- Beveled edges provide easy access to channels
Specifications
15 | |
Natural | |
Microscope Slide | |
Non-sterile |
Not Autoclavable | |
153 x 102 x 21 mm | |
HDPE |