missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate

Color:Clear; Sterility:Sterile; Lid:With Lid

Catalog No. FB012931


Explore available promotions

Description

Description

  • Uniform well volume to ensure equal growth surface area
  • Well surface smooth, free from striation to maximize usable growth area
  • Raised rims on wells with uniform rings on lid to reduce evaporation
  • Single position lid reduces risks of cross-contamination, handling mistakes
  • Alphanumeric code on wells for easy identification
  • Suitable for all common instruments, automation
  • Sterilized by gamma irradiation
  • Non-pyrogenic

Warranty

Three years

Specifications

Specifications

Alphanumeric
Standard
0.33 cm2
Clear
0.39 mL
Sterile
Flat Bottom (F-well)
100 Plates
Raised rims with condensation rings
CE
With Lid
8 x 12
Virgin Polystyrene
96
Cell Culture
Tissue culture
Yes
SDS
Documents

Documents

Promotions

Promotions