missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet
Get more space to store hazardous material in your work environment. Justrite Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet smaller sized cabinets fit nearly anywhere.
$2259.46 - $3402.24
Specifications
Depth (English) Interior | 14.56 in. |
---|---|
Depth (Metric) Exterior | 45.7 cm |
Depth (Metric) Interior | 37 cm |
Color | Blue |
Depth (English) Exterior | 18 in. |
Alert:
These cabinets are not recommended for storage of phenol, nitric acid or sulfuric acid.
Catalog Number | Mfr. No. | Load Bearing Capacity | Certifications/Compliance | Dimensions (D x W x H) Interior | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Load Bearing Capacity | Certifications/Compliance | Dimensions (D x W x H) Interior | Price | Quantity | |||
19-159-719
|
Justrite™
8912022 |
12 gal. (45 L) | NFPA, OSHA | 14.56 x 20 x 30.5 in. (37 x 50.8 x 77.5 cm) |
Each for $3,402.24
|
|
|||
19-413-822
|
Justrite™
891502 |
15 gal. (57 L) | FM, NFPA, OSHA | 14.56 x 20 x 39.6 in. (37 x 50.8 x 100.6 cm) |
Each for $2,259.46
|
|
|||
Description
- Flame-coated thermoplastic liner
- 100% seamless liner on all interior walls, ceiling, sump, inside doors and shelves
- Seam-free surface
- Non-porous surface resists staining and cleans-up easily
- Dual-benefits/Multi-use - complete protection from harsh acids and corrosive chemicals
- Blue color
- Features double-wall, 18-gauge (1mm) welded steel construction, 2 in. (51mm) liquid-tight containment sump, dual vents
- Adjustable shelves, three-point stainless steel bullet self-latching doors with ULoc™ padlockable handle and Haz-Alert™ reflective warning labels
- ChemCor™ liner interior
Compliance
Compliance: Retains FM Approval & Complies with OSHA regulations and the International Fire Code of NFPA 1 Uniform Fire Code. Only NFPA/OSHA for 19159712/19413871.
Specifications
14.56 in. | |
37 cm | |
18 in. | |
23.25 in. | |
Flammables | |
1 | |
59.05 cm |
45.7 cm | |
Blue | |
Steel (CR) | |
Manual-close | |
1 | |
20 in. | |
50.8 cm |